(Difluoromethyl)benzene ≥98.0% (by GC)

Supplier: TCI America
Danger

D4915-1G D4915-5G
TCD4915-1GEA 139 USD
TCD4915-1G TCD4915-5G
(Difluoromethyl)benzene ≥98.0% (by GC)
(Difluoromethyl)benzene
Formula: C₇H₆F₂
Boiling Pt: 135°C
Flash Pt: 35°C
MDL Number: MFCD03788488
CAS Number: 455-31-2
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR