3-(Perfluoro-n-hexyl)-1,2-propenoxide ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoroheptyloxirane, 3-Perfluorohexyl-1,2-epoxypropane

T3365-25G T3365-5G
76220-580EA 604 USD
76220-580 76220-582
3-(Perfluoro-n-hexyl)-1,2-propenoxide ≥98.0% (by GC)
3-(Perfluoro-n-hexyl)-1,2-propenoxide
Formula: C₉H₅F₁₃O
Boiling Pt: 154°C
Flash Pt: 82°C
MDL Number: MFCD00042356
CAS Number: 38565-52-5
EINECS: 254-004-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR