Difluoromethyl-2,2,3,3-tetrafluoropropyl ether ≥98.0% (by GC)

Supplier: TCI America
Danger

D4472-1G D4472-5G
TCD4472-1GEA 80 USD
TCD4472-1G TCD4472-5G
Difluoromethyl-2,2,3,3-tetrafluoropropyl ether ≥98.0% (by GC)
Difluoromethyl-2,2,3,3-tetrafluoropropyl ether
Formula: C₄H₄F₆O
MW: 182.07 g/mol
Boiling Pt: 76°C
MDL Number: MFCD00236119
CAS Number: 35042-99-0
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR