4-(4-Chlorophenyl)-5-methyl-1,3-thiazol-2-amine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-(4-Chlorophenyl)-5-methylthiazol-2-amine, 2-Amino-4-(4-chlorophenyl)-5-methylthiazole

A3066-1G A3066-5G
TCA3066-1GEA 78 USD
TCA3066-1G TCA3066-5G
4-(4-Chlorophenyl)-5-methyl-1,3-thiazol-2-amine ≥98.0% (by GC)
4-(4-Chlorophenyl)-5-methyl-1,3-thiazol-2-amine
Formula: C₁₀H₉ClN₂S
MW: 224.71 g/mol
MDL Number: MFCD00731832
CAS Number: 82632-77-7

Order Now

Specification Test Results

Appearance White to Yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 145.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR