3,6-Dibromo-2,7-dihydroxynaphthalene ≥98.0% (by GC)

Supplier: TCI America

D4669-1G D4669-5G
TCD4669-1GEA 153 USD
TCD4669-1G TCD4669-5G
3,6-Dibromo-2,7-dihydroxynaphthalene ≥98.0% (by GC)
3,6-Dibromo-2,7-dihydroxynaphthalene
Formula: C₁₀H₆Br₂O₂
MDL Number: MFCD00277643
CAS Number: 96965-79-6

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR