5-Iodopyridin-2-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Hydroxy-5-iodopyridine, 5-Iodopyridin-2(1H)-one

H1466-1G H1466-5G
TCH1466-1GEA 52.3 USD
TCH1466-1G TCH1466-5G
5-Iodopyridin-2-ol ≥98.0% (by GC, titration analysis)
5-Iodopyridin-2-ol
Formula: C₅H₄INO
MDL Number: MFCD00234058
CAS Number: 13472-79-2

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 189.0 to 194.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR