Perfluoro(2-methyl-3-pentanone) ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone, Perfluoroethyl Perfluoroisopropyl ketone

N1038-100G N1038-25G
TCN1038-100GEA 420 USD
TCN1038-100G TCN1038-25G
Perfluoro(2-methyl-3-pentanone) ≥98.0% (by GC)
Perfluoro(2-methyl-3-pentanone)
Formula: C₆F₁₂O
MW: 316.05 g/mol
MDL Number: MFCD04038343
CAS Number: 756-13-8
EINECS: 436-710-6

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR