4-Methylumbelliferyl-Β-D-cellobiopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
M3028-100MG
M3028-500MG
TCM3028-100MGEA
133
USD
TCM3028-100MG
TCM3028-500MG
4-Methylumbelliferyl-Β-D-cellobiopyranoside ≥98.0% (by HPLC)
4-Methylumbelliferyl-Β-D-cellobiopyranoside
Formula:
C₂₂H₂₈O₁₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00063279 CAS Number: 72626-61-0 EINECS: 276-744-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -92.0 to -97.0 deg (C=0.2, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...