Methyl-3,3,3-trifluoropropanate ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
M2783-1G
M2783-5G
TCM2783-1GEA
95
USD
TCM2783-1G
TCM2783-5G
Methyl-3,3,3-trifluoropropanate ≥98.0% (by GC)
Methyl-3,3,3-trifluoropropanate
Formula:
C₄H₅F₃O₂ Boiling Pt: 96°C |
MDL Number:
MFCD02262206 CAS Number: 18830-44-9 UN: 3272 ADR: 3,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...