3',5'-Dichloro-4'-Hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D5020-1G
D5020-5G
TCD5020-1GEA
130
USD
TCD5020-1G
TCD5020-5G
3',5'-Dichloro-4'-Hydroxyacetophenone ≥98.0% (by GC, titration analysis)
3',5'-Dichloro-4'-Hydroxyacetophenone
MDL Number:
MFCD00016421 CAS Number: 17044-70-1 EINECS: 241-113-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 162.0 to 166.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...