(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B4418-25G B4418-5G
TCB4418-25GEA 398 USD
TCB4418-25G TCB4418-5G
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine ≥98.0% (by HPLC, total nitrogen)
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine
Formula: C₃₆H₅₄N₂O₂
MDL Number: MFCD00191800
CAS Number: 135616-40-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 206.0 to 210.0 deg-C
Specific rotation [a]20/D -310 to -330 deg (C=1,CH2Cl2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR