Hydroxocobalamin acetate salt ≥90.0% (by HPLC)

Supplier: TCI America

H1542-1G H1542-5G
TCH1542-1GEA 215.66 USD
TCH1542-1G TCH1542-5G
Hydroxocobalamin acetate salt ≥90.0% (by HPLC)
Hydroxocobalamin acetate salt
Formula: C₆₄H₉₂CoN₁₃O₁₆P
Storage Temperature: Refrigerator
MDL Number: MFCD00082532
CAS Number: 22465-48-1
EINECS: 245-019-7
Merck Index: 14,04809

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (Chelometric Titration) min. 95.0 % (calcd.on dried substance)
Drying loss max. 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR