4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside, 4-Methylumbelliferyl N-Acetyl-beta-D-glucosaminide

M3030-1G M3030-250MG
TCM3030-1GEA 319 USD
TCM3030-1G TCM3030-250MG
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC, total nitrogen)
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside
Formula: C₁₈H₂₁NO₈
MW: 379.365 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065481
CAS Number: 37067-30-4
EINECS: 253-333-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -29.0 to -33.0 deg (C=1, Pyridine)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR