trans-N-Fmoc-4-tert-butoxy-L-proline ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-Fmoc-4-trans-(tert-butoxy)-L-proline, Fmoc-Hyp(tBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-trans-(tert-butoxy)-L-proline, (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxy)pyrrolidine-2-carboxylic acid

F1095-1G F1095-5G
TCF1095-1GEA 34 USD
TCF1095-1G TCF1095-5G
trans-N-Fmoc-4-tert-butoxy-L-proline ≥98.0% (by HPLC)
trans-N-Fmoc-4-tert-butoxy-L-proline
Formula: C₂₄H₂₇NO₅
MW: 409.482 g/mol
MDL Number: MFCD00151930
CAS Number: 122996-47-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %
Specific rotation [a]20/D -23.0 to -28.0 deg (C=1, MeOH)
Drying loss max. 2.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR