2-Methylallylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 3-Amino-2-methyl-1-propene

M2726-1ML M2726-5ML
TCM2726-1MLEA 78 USD
TCM2726-1ML TCM2726-5ML
2-Methylallylamine ≥98.0% (by GC, titration analysis)
2-Methylallylamine
Formula: C₄H₉N
MW: 71.123 g/mol
Boiling Pt: 81°C
MDL Number: MFCD00053646
CAS Number: 2878-14-0
EINECS: 220-724-2
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR