4'-Chloro-2,2':6',2''-terpyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
C3187-1G
C3187-200MG
TCC3187-1GEA
186
USD
TCC3187-1G
TCC3187-200MG
4'-Chloro-2,2':6',2''-terpyridine ≥98.0% (by GC, titration analysis)
4'-Chloro-2,2':6',2''-terpyridine
Formula:
C₁₅H₁₀ClN₃ MW: 267.716 g/mol |
MDL Number:
MFCD00191930 CAS Number: 128143-89-5 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...