1-Adamantyl isocyanate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
A1706-1G
A1706-5G
TCA1706-1GEA
185
USD
TCA1706-1G
TCA1706-5G
1-Adamantyl isocyanate ≥98.0% (by GC)
1-Adamantyl isocyanate
Formula:
C₁₁H₁₅NO |
MDL Number:
MFCD00134405 CAS Number: 4411-25-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 145.0 to 149.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...