4-Hydroxy-7-methoxycoumarin ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Hydroxy-7-methoxy-2H-chromen-2-one

H1420-1G H1420-5G
TCH1420-1GEA 96 USD
TCH1420-1G TCH1420-5G
4-Hydroxy-7-methoxycoumarin ≥98.0% (by GC, titration analysis)
4-Hydroxy-7-methoxycoumarin
Formula: C₁₀H₈O₄
MW: 192.17 g/mol
MDL Number: MFCD00673700
CAS Number: 17575-15-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR