Triisopropanolamine cyclic borate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
|
Warning
|
Synonyms:
3,7,10-Trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo[3.3.3]undecane, Triisopropanolamine Borate, Triisopropanolamine Cyclic Borate
T2943-25G
T2943-5G
TCT2943-25GEA
252
USD
TCT2943-25G
TCT2943-5G
Triisopropanolamine cyclic borate ≥98.0% (by total nitrogen basis)
Triisopropanolamine cyclic borate
Formula:
C₉H₁₈BNO₃ Boiling Pt: 135°C |
MDL Number:
MFCD03791298 CAS Number: 101-00-8 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...