N-(Propargyloxy)phthalimide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-(Prop-2-yn-1-yloxy)isoindoline-1,3-dione
P2342-1G
P2342-5G
TCP2342-1GEA
45
USD
TCP2342-1G
TCP2342-5G
N-(Propargyloxy)phthalimide ≥98.0% (by GC)
N-(Propargyloxy)phthalimide
Formula:
C₁₁H₇NO₃ MW: 201.181 g/mol |
MDL Number:
MFCD00005890 CAS Number: 4616-63-1 EINECS: 225-024-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...