4,6-Dibromodibenzothiophene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4,6-Dibromodibenzo[b,d]thiophene

D4822-1G D4822-200MG
TCD4822-1GEA 147 USD
TCD4822-1G TCD4822-200MG
4,6-Dibromodibenzothiophene ≥98.0% (by GC)
4,6-Dibromodibenzothiophene
Formula: C₁₂H₆Br₂S
MW: 342.048 g/mol
MDL Number: MFCD18451473
CAS Number: 669773-34-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 184.0 to 188.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR