4-tert-Butylphenyl glycidyl ether ≥98.0% (by GC)

Supplier: TCI America
Warning

B4831-100G B4831-25G
TCB4831-100GEA 333 USD
TCB4831-100G TCB4831-25G
4-tert-Butylphenyl glycidyl ether ≥98.0% (by GC)
4-tert-Butylphenyl glycidyl ether
Formula: C₁₃H₁₈O₂
Boiling Pt: 294°C
Flash Pt: 102°C
MDL Number: MFCD00005136
CAS Number: 3101-60-8
EINECS: 221-453-2

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR