N-Methyl-3-nitroaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M3042-1G M3042-5G
TCM3042-1GEA 191 USD
TCM3042-1G TCM3042-5G
N-Methyl-3-nitroaniline ≥98.0% (by GC, titration analysis)
N-Methyl-3-nitroaniline
Formula: C₇H₈N₂O₂
MW: 152.153 g/mol
MDL Number: MFCD00963641
CAS Number: 619-26-1

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR