BAY 11-7085 ≥97.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: (E)-3-(4-tert-Butylphenylsulphonyl)acrylonitrile, (E)-3-((4-(tert-Butyl)phenyl)sulphonyl)acrylonitrile

B4845-100MG B4845-10MG
TCB4845-100MGEA 504.2 USD
TCB4845-100MG TCB4845-10MG
BAY 11-7085 ≥97.0% (by total nitrogen basis)
BAY 11-7085
Formula: C₁₃H₁₅NO₂S
MW: 249.328 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862602
CAS Number: 196309-76-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR