Nimesulide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: N-(4-Nitro-2-phenoxyphenyl)methanesulphonamide

N0984-1G N0984-5G
TCN0984-1GEA 21 USD
TCN0984-1G TCN0984-5G
Nimesulide ≥98.0% (by HPLC, titration analysis)
Nimesulide
Formula: C₁₃H₁₂N₂O₅S
MW: 308.308 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00079470
CAS Number: 51803-78-2
EINECS: 257-431-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,06548

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 148.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR