((Difluoromethyl)sulfonyl)benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Difluoromethyl phenyl sulphone

D4764-1G D4764-5G
TCD4764-1GEA 72 USD
TCD4764-1G TCD4764-5G
((Difluoromethyl)sulfonyl)benzene ≥98.0% (by GC)
((Difluoromethyl)sulfonyl)benzene
Formula: C₇H₆F₂O₂S
MW: 192.18 g/mol
MDL Number: MFCD01050170
CAS Number: 1535-65-5

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Refractive index n20/D 1.5000 to 1.5040


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR