4-Bromo-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Bromo-9,9-dimethylfluorene
B4661-1G
B4661-200MG
TCB4661-1GEA
68
USD
TCB4661-1G
TCB4661-200MG
4-Bromo-9,9-dimethyl-9H-fluorene ≥98.0% (by GC)
4-Bromo-9,9-dimethyl-9H-fluorene
Formula:
C₁₅H₁₃Br MW: 273.173 g/mol |
MDL Number:
MFCD23099415 CAS Number: 942615-32-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...