2-Hydroxyisonicotinic acid ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid

H1336-25G H1336-5G
TCH1336-25GEA 269 USD
TCH1336-25G TCH1336-5G
2-Hydroxyisonicotinic acid ≥98.0% (by GC)
2-Hydroxyisonicotinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 328°C
MDL Number: MFCD01318566
CAS Number: 22282-72-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR