3-Fluoro-4'-n-propylbiphenyl-4-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-Fluoro-4'-propyl-4-biphenylboronic acid

F0942-1G F0942-5G
TCF0942-1GEA 69 USD
TCF0942-1G TCF0942-5G
3-Fluoro-4'-n-propylbiphenyl-4-boronic acid (contains varying amounts of Anhydride)
3-Fluoro-4'-n-propylbiphenyl-4-boronic acid
Formula: C₁₅H₁₆BFO₂
MW: 258.099 g/mol
MDL Number: MFCD10687198
CAS Number: 909709-42-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR