Isoindoline hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

I0958-1G I0958-5G
TCI0958-1GEA 22 USD
TCI0958-1G TCI0958-5G
Isoindoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Isoindoline hydrochloride
Formula: C₈H₁₀ClN
MW: 155.625 g/mol
MDL Number: MFCD03502377
CAS Number: 32372-82-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 259.0 to 263.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR