trans-4-(4-n-Propylcyclohexyl)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-(trans-4-Propylcyclohexyl)benzoic acid, 4-(trans-4-n-Propylcyclohexyl)benzoic acid

P2438-25G P2438-5G
TCP2438-25GEA 129 USD
TCP2438-25G TCP2438-5G
trans-4-(4-n-Propylcyclohexyl)benzoic acid ≥98.0% (by GC, titration analysis)
trans-4-(4-n-Propylcyclohexyl)benzoic acid
Formula: C₁₆H₂₂O₂
Melting Pt: 206°C
MDL Number: MFCD06658177
CAS Number: 65355-29-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Mesomorphic range 214.0 to 262.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR