trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-n-propylbicyclohexyl ≥98.0% (by GC)
Supplier: TCI America
E1157-1G
E1157-5G
TCE1157-1GEA
55.4
USD
TCE1157-1G
TCE1157-5G
trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-n-propylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-n-propylbicyclohexyl
Formula:
C₂₃H₃₄F₂O |
MDL Number:
MFCD18433524 CAS Number: 123560-48-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Mesomorphic range | 78.0 to 84.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...