4-Methoxybenzenediazonium tetrafluoroborate ≥98.0% (by HPLC)

Supplier: TCI America
Danger

M2636-25G M2636-5G
TCM2636-25GEA 532 USD
TCM2636-25G TCM2636-5G
4-Methoxybenzenediazonium tetrafluoroborate ≥98.0% (by HPLC)
4-Methoxybenzenediazonium tetrafluoroborate
Formula: C₇H₇BF₄N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00011897
CAS Number: 459-64-3
EINECS: 207-296-2
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR