Potassium tricyanomethanide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
P2538-1G
P2538-5G
TCP2538-1GEA
153
USD
TCP2538-1G
TCP2538-5G
Potassium tricyanomethanide ≥98.0% (by titrimetric analysis)
Potassium tricyanomethanide
Formula:
C₄KN₃ Melting Pt: 298°C |
MDL Number:
MFCD00058850 CAS Number: 34171-69-2 UN: 3439 ADR: 6.1,II |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...