Sodium-tert-pentyloxide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

S0950-100G S0950-25G
TCS0950-100GEA 58 USD
TCS0950-100G TCS0950-25G
Sodium-tert-pentyloxide ≥97.0% (by titrimetric analysis)
Sodium-tert-pentyloxide
Formula: C₅H₁₁NaO
MDL Number: MFCD00064245
CAS Number: 14593-46-5
EINECS: 238-639-4
UN: 3206
ADR: 4.2,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR