Phenethyltrimethoxysilane ≥95.0% (by GC)

Supplier: TCI America
Warning

T3542-100G T3542-25G
TCT3542-100GEA 315 USD
TCT3542-100G TCT3542-25G
Phenethyltrimethoxysilane ≥95.0% (by GC)
Phenethyltrimethoxysilane
Formula: C₁₁H₁₈O₃Si
Boiling Pt: 122°C
Flash Pt: 109°C
MDL Number: MFCD00053852
CAS Number: 49539-88-0
EINECS: 256-363-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Red clear liquid
Purity (GC) min. 95.0 % (total of isomer)
Specific gravity (20/20) 1.0390 to 1.0440
Refractive index n20/D 1.4750 to 1.4800


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR