trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Fluoro-4'-(trans-4-propylcyclohexyl)biphenyl, 4-Fluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

F1103-1G F1103-5G
TCF1103-1GEA 41 USD
TCF1103-1G TCF1103-5G
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl ≥98.0% (by GC)
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl
Formula: C₂₁H₂₅F
MW: 296.429 g/mol
MDL Number: MFCD11053438
CAS Number: 87260-24-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 104 to 160 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR