N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B4810-25G
B4810-5G
TCB4810-25GEA
73.2
USD
TCB4810-25G
TCB4810-5G
N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline ≥98.0% (by HPLC, total nitrogen)
N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline
Formula:
C₁₈H₁₉FN₂O₃ |
MDL Number:
MFCD06658240 CAS Number: 168828-81-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...