1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥98.0% (by GC)

Supplier: TCI America

E1408-25G E1408-5G
TCE1408-25GEA 342 USD
TCE1408-25G TCE1408-5G
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥98.0% (by GC)
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene
Formula: C₁₇H₁₆
MDL Number: MFCD04038793
CAS Number: 22692-80-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR