Poly(3-hexylthiophene-2,5-diyl) regioregular
Supplier: TCI America
|
Warning
|
Synonyms:
P3HT, Poly(3-hexylthiophene), Poly[3-hexylthiophene-2,5-diyl]
P2513-100MG
P2513-500MG
TCP2513-100MGEA
264
USD
TCP2513-100MG
TCP2513-500MG
Poly(3-hexylthiophene-2,5-diyl) regioregular
Poly(3-hexylthiophene-2,5-diyl)
MW:
196.352 g/mol |
MDL Number:
MFCD00217686 CAS Number: 104934-50-1 |
Specification Test Results
Hole Mobility (mu FET) | min 0.10cm2/Vs (OTS Si/SiO2 substrate) |
Palladium | max. 100 ppm |
Molecular weight of number average | 27,000 to 45,000 |
Stereoregularity | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...