6-Chloro-7-iodo-7-deazapurine ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine, 6-Chloro-7-iodo-deazapurine

C3130-1G C3130-5G
TCC3130-1GEA 31 USD
TCC3130-1G TCC3130-5G
6-Chloro-7-iodo-7-deazapurine ≥95.0% (by GC, titration analysis)
6-Chloro-7-iodo-7-deazapurine
Formula: C₆H₃ClIN₃
MW: 279.47 g/mol
MDL Number: MFCD09263258
CAS Number: 123148-78-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 200.0 to 204.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR