Ruthenium(III) chloride oxide

Supplier: TCI America

Synonyms: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

R0204-1G R0204-5G
TCR0204-1GEA 489.6 USD
TCR0204-1G TCR0204-5G
Ruthenium(III) chloride oxide
Ruthenium(III) chloride oxide
Formula: H₄₂Cl₆N₁₄O₂Ru₃
MW: 786.354 g/mol
MDL Number: MFCD00011479
CAS Number: 11103-72-3

Order Now

Specification Test Results

Appearance Gray to Dark purple to Black powder to crystal
Lambda max. 532.0 to 536.0 nm (0.1mol/L AcONH4 sol.)
Absorbance (E1%1cm) min. 250 (0.1mol/L AcONH4 sol., 532.0 to 536.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR