1-(Phenylsulfonyl)pyrrole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
P2095-1G
P2095-5G
TCP2095-1GEA
17
USD
TCP2095-1G
TCP2095-5G
1-(Phenylsulfonyl)pyrrole ≥98.0% (by GC)
1-(Phenylsulfonyl)pyrrole
Formula:
C₁₀H₉NO₂S MW: 207.247 g/mol |
MDL Number:
MFCD00067739 CAS Number: 16851-82-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...