Biotin-LC-LC-NHS ≥90.0% (by HPLC)

Supplier: TCI America

Synonyms: N-Succinimidyl N-[6-(Biotinamido)hexanoyl]-6-aminohexanoate, Biotin-LC-LC-NHS (2mg*5), 2,5-Dioxopyrrolidin-1-yl 6-(6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanamido)hexanoate, N-[6-(Biotinamido)hexanoyl]-6-aminohexanoic acid N-Succinimidyl ester

S0956-100MG S0956-25MG
TCS0956-100MGEA 462 USD
TCS0956-100MG TCS0956-25MG
Biotin-LC-LC-NHS ≥90.0% (by HPLC)
Biotin-LC-LC-NHS
Formula: C₂₆H₄₁N₅O₇S
Storage Temperature: Freezer
MDL Number: MFCD00467154
CAS Number: 89889-52-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 90.0 area%
Elemental analysis (Nitrogen) 11.70 to 13.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR