Imperatorin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 9-((3-Methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one

I0904-100MG I0904-25MG
TCI0904-100MGEA 206 USD
TCI0904-100MG TCI0904-25MG
Imperatorin ≥98.0% (by HPLC)
Imperatorin
Formula: C₁₆H₁₄O₄
MW: 270.284 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00016881
CAS Number: 482-44-0
EINECS: 207-581-1
Merck Index: 14,04925

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Melting point 97.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR