1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 7-Chloro-4-[4-(3-[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propyl)-1-piperazinyl]quinoline tetraphosphate tetrahydrate, UNII-854D7K8LXB, Piperaquine tetraphosphate tetrahydrate

P1987-25G P1987-5G
TCP1987-25GEA 206 USD
TCP1987-25G TCP1987-5G
1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate ≥98.0% (by HPLC, total nitrogen)
1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate
Formula: C₂₉H₃₂Cl₂N₆·4H₃PO₄.4H₂O
MW: 999.552 g/mol
Melting Pt: 252°C
MDL Number: MFCD11870901
CAS Number: 915967-82-7
New Product

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Water 5.8 to 8.6 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR