Tyrphostin 1 ≥98.0% (by GC)

Supplier: TCI America
Danger

T3074-1G T3074-200MG
TCT3074-1GEA 559.7 USD
TCT3074-1G TCT3074-200MG
Tyrphostin 1 ≥98.0% (by GC)
Tyrphostin 1
Formula: C₁₁H₈N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00019787
CAS Number: 2826-26-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR