Cefmetazole sodium ≥92.0% (by HPLC)

Supplier: TCI America
Warning

C3029-1G C3029-250MG
TCC3029-1GEA 239 USD
TCC3029-1G TCC3029-250MG
Cefmetazole sodium ≥92.0% (by HPLC)
Cefmetazole sodium
Formula: C₁₅H₁₆N₇NaO₅S₃
MDL Number: MFCD00214260
CAS Number: 56796-39-5
Merck Index: 14,01926

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 92.0 area%
Purity (Ion exchange titration) min. 97.0 %
Specific rotation [a]20/D +73.0 to +85.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR