dibenzo[b,d]furan-2-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

D4759-1G D4759-5G
TCD4759-1GEA 134 USD
TCD4759-1G TCD4759-5G
dibenzo[b,d]furan-2-carbaldehyde ≥98.0% (by GC)
dibenzo[b,d]furan-2-carbaldehyde
Formula: C₁₃H₈O₂
MDL Number: MFCD00136272
CAS Number: 5397-82-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR