9-Bromo-1-nonene ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
B3256-1G
B3256-5G
TCB3256-1GEA
36
USD
TCB3256-1G
TCB3256-5G
9-Bromo-1-nonene ≥95.0% (by GC)
9-Bromo-1-nonene
Formula:
C₉H₁₇Br Boiling Pt: 101°C Storage Temperature: Refrigerator |
MDL Number:
MFCD09037826 CAS Number: 89359-54-6 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...